COMH4211D3DD Combi-Blocks, Inc.

Sulfobromophthalein sodium salt hydrate

  • MDL number MFCD00150017
  • Cas Number : 71-67-0
  • Molecular Weight : 856.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7134

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7134
Availability details for
Quantity
  • Availability
  • Add to Cart