COMH4211D436 Combi-Blocks, Inc.

Methyl 2-amino-2-(thiophen-2-yl)acetate hydrochloride

  • MDL number MFCD04039779
  • Cas Number : 132289-66-8
  • Molecular Weight : 207.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-9340

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-9340
Availability details for
Quantity
  • Availability
  • Add to Cart