COMH4211D43E Combi-Blocks, Inc.

Sodium phenoxide

  • MDL number MFCD00013134
  • Cas Number : 139-02-6
  • Molecular Weight : 116.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-9500

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-9500
Availability details for
Quantity
  • Availability
  • Add to Cart