COMH4211D451 Combi-Blocks, Inc.

Bis(p-tolyl)phosphine oxide

  • MDL number MFCD01445489
  • Cas Number : 2409-61-2
  • Molecular Weight : 230.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-9973

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-9973
Availability details for
Quantity
  • Availability
  • Add to Cart