COMH4211D458 Combi-Blocks, Inc.

(S)-(+)-2-Hydroxy-2-phenylpropionic acid

  • MDL number MFCD00067700
  • Cas Number : 13113-71-8
  • Molecular Weight : 166.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0149

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0149
Availability details for
Quantity
  • Availability
  • Add to Cart