COMH4211D45D Combi-Blocks, Inc.

5-Methyl-2-phenyl-1h-benzoimidazole

  • MDL number MFCD00458486
  • Cas Number : 2963-65-7
  • Molecular Weight : 208.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0765

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0765
Availability details for
Quantity
  • Availability
  • Add to Cart