COMH4211D469 Combi-Blocks, Inc.

3-(2-Fluoro-5-methylphenyl)propionic acid

  • MDL number MFCD09025395
  • Cas Number : 881189-60-2
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1706

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1706
Availability details for
Quantity
  • Availability
  • Add to Cart