COMH4211D46B Combi-Blocks, Inc.

Sodium tetrabromoaurate(iii) hydrate

  • MDL number MFCD00274624
  • Cas Number : 10378-49-1
  • Molecular Weight : 557.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1773

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1773
Availability details for
Quantity
  • Availability
  • Add to Cart