COMH4211D476 Combi-Blocks, Inc.

2-(2-tert-Butyl-1,3-thiazol-4-yl)acetic acid

  • MDL number MFCD11180931
  • Cas Number : 1094411-96-7
  • Molecular Weight : 199.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-2598

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-2598
Availability details for
Quantity
  • Availability
  • Add to Cart