COMH4211D47C Combi-Blocks, Inc.

3-(Boc-aminomethyl)-4-fluorobenzeneboronic acid pinacol ester

  • MDL number MFCD22494729
  • Cas Number : 1035391-48-0
  • Molecular Weight : 351.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-2882

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-2882
Availability details for
Quantity
  • Availability
  • Add to Cart