COMH4211D485 Combi-Blocks, Inc.

4-(2-Chloroquinazolin-4-yl)morpholine

  • MDL number MFCD03083869
  • Cas Number : 39213-05-3
  • Molecular Weight : 249.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-3270

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-3270
Availability details for
Quantity
  • Availability
  • Add to Cart