COMH4211D499 Combi-Blocks, Inc.

4-Ethoxy-2-fluorobenzonitrile

  • MDL number MFCD13196591
  • Cas Number : 186191-34-4
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4481

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4481
Availability details for
Quantity
  • Availability
  • Add to Cart