COMH4211D4C0 Combi-Blocks, Inc.

4-Fluoro-2-methyl-thiobenzamide

  • MDL number MFCD09757564
  • Cas Number : 1256482-73-1
  • Molecular Weight : 169.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7436

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7436
Availability details for
Quantity
  • Availability
  • Add to Cart