COMH4211D4DA Combi-Blocks, Inc.

3-Chloro-4-(trifluoromethyl)benzamide

  • MDL number MFCD11519326
  • Cas Number : 1092460-78-0
  • Molecular Weight : 223.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9195

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9195
Availability details for
Quantity
  • Availability
  • Add to Cart