COMH4211D4DE Combi-Blocks, Inc.

4-(3-Methyl-1,2,4-oxadiazol-5-yl)piperidine hydrochloride

  • MDL number MFCD12912978
  • Cas Number : 1181540-59-9
  • Molecular Weight : 203.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9483

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9483
Availability details for
Quantity
  • Availability
  • Add to Cart