COMH4211D4E2 Combi-Blocks, Inc.

4-Chloro-n-(4-methoxyphenyl)butyramide

  • MDL number MFCD03367954
  • Molecular Weight : 227.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9777

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9777
Availability details for
Quantity
  • Availability
  • Add to Cart