COMH4211D542 Combi-Blocks, Inc.

4-(Bromomethyl)quinoline

  • MDL number MFCD09953523
  • Cas Number : 5632-16-6
  • Molecular Weight : 222.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3621

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3621
Availability details for
Quantity
  • Availability
  • Add to Cart