COMH4211D547 Combi-Blocks, Inc.

(Z,1R,8S,9S)-Ethyl bicyclo[6.1.0]non-4-ene-9-carboxylate

  • MDL number MFCD20264901
  • Cas Number : 1263291-39-9
  • Molecular Weight : 194.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3891

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3891
Availability details for
Quantity
  • Availability
  • Add to Cart