COMH4211D553 Combi-Blocks, Inc.

Tropine-3-mesylate

  • MDL number MFCD12965025
  • Cas Number : 35130-97-3
  • Molecular Weight : 219.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4227

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4227
Availability details for
Quantity
  • Availability
  • Add to Cart