COMH4211DD2F Combi-Blocks, Inc.

Iodomesitylene diacetate

  • MDL number MFCD00674196
  • Cas Number : 33035-41-5
  • Molecular Weight : 362.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6507

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6507
Availability details for
Quantity
  • Availability
  • Add to Cart