COMH4211DD50 Combi-Blocks, Inc.

(1-Benzylazetidin-2-yl)methanol

  • MDL number MFCD11845667
  • Cas Number : 31247-34-4
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6774

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6774
Availability details for
Quantity
  • Availability
  • Add to Cart