COMH4211DD5A Combi-Blocks, Inc.

3-Nitrobenzoyl isothiocyanate

  • MDL number MFCD00041247
  • Cas Number : 78225-78-2
  • Molecular Weight : 208.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 60
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-7261

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-7261
Availability details for
Quantity
  • Availability
  • Add to Cart