COMH4211DDC4 Combi-Blocks, Inc.

Atractyloside potassium salt

  • MDL number MFCD00078810
  • Cas Number : 102130-43-8
  • Molecular Weight : 803.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0356

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0356
Availability details for
Quantity
  • Availability
  • Add to Cart