COMH4211DDD7 Combi-Blocks, Inc.

4H-Thieno[3,2-b]pyrrole-5-carboxylic acid hydrazide

  • MDL number MFCD04208027
  • Cas Number : 119448-43-0
  • Molecular Weight : 181.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0565

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0565
Availability details for
Quantity
  • Availability
  • Add to Cart