COMH4211DDE9 Combi-Blocks, Inc.

(S)-2-(((9H-Fluoren-9-yl)methoxy)carbonylamino)-2-phenylpropanoic acid

  • MDL number MFCD11052918
  • Cas Number : 881921-11-5
  • Molecular Weight : 387.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0670

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0670
Availability details for
Quantity
  • Availability
  • Add to Cart