COMH4211DDF0 Combi-Blocks, Inc.

Methyl 3,4-difluorobenzoylformate

  • MDL number MFCD01631252
  • Cas Number : 220239-71-4
  • Molecular Weight : 200.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0750

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0750
Availability details for
Quantity
  • Availability
  • Add to Cart