COMH4211DDFB Combi-Blocks, Inc.

Ethyl 5-(chlorosulfonyl)-3-furoate

  • MDL number MFCD11109333
  • Cas Number : 256373-91-8
  • Molecular Weight : 238.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0844

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0844
Availability details for
Quantity
  • Availability
  • Add to Cart