COMH4211DE2A Combi-Blocks, Inc.

T-Butyloxycarbonyll-peg5-1-o-(b-cyanoethyl-n,n-diisopropyl)phosphoramidite

  • MDL number MFCD29037306
  • Cas Number : 1358906-70-3
  • Molecular Weight : 522.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-1226

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-1226
Availability details for
Quantity
  • Availability
  • Add to Cart