COMH4211DE2F Combi-Blocks, Inc.

[(3-Bromophenyl)methyl](cyclopropylmethyl)amine

  • MDL number MFCD11140259
  • Cas Number : 1019506-79-6
  • Molecular Weight : 240.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-1270

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-1270
Availability details for
Quantity
  • Availability
  • Add to Cart