COMH4211DE47 Combi-Blocks, Inc.

2-Fluoro-5-methylphenylacetonitrile

  • MDL number MFCD07772783
  • Cas Number : 945541-97-9
  • Molecular Weight : 149.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-1517

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-1517
Availability details for
Quantity
  • Availability
  • Add to Cart