COMH4211DE52 Combi-Blocks, Inc.

Sesamolin

  • MDL number MFCD04112844
  • Cas Number : 526-07-8
  • Molecular Weight : 370.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-1636

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-1636
Availability details for
Quantity
  • Availability
  • Add to Cart