COMH4211DEC4 Combi-Blocks, Inc.

3,4-Dimethylthieno[2,3-b]thiophene-2-carbaldehyde

  • MDL number MFCD00052282
  • Cas Number : 159709-36-1
  • Molecular Weight : 196.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-2663

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-2663
Availability details for
Quantity
  • Availability
  • Add to Cart