COMH4211DED9 Combi-Blocks, Inc.

1-(3-(4-Bromophenylsulfonyl)propyl)pyrrolidine

  • MDL number MFCD12024825
  • Cas Number : 867333-33-3
  • Molecular Weight : 332.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-2827

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-2827
Availability details for
Quantity
  • Availability
  • Add to Cart