COMH4211DEF8 Combi-Blocks, Inc.

5-Chloro-2-(isopropylamino)aniline

  • MDL number MFCD00084956
  • Cas Number : 89659-68-7
  • Molecular Weight : 184.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-3110

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-3110
Availability details for
Quantity
  • Availability
  • Add to Cart