COMH4211DF22 Combi-Blocks, Inc.

(1R*,2S*,4S*)-7-Aza-bicyclo[2.2.1]heptane-2,7-dicarboxylic acid 7-tert-butyl ester

  • MDL number MFCD12198742
  • Cas Number : 500556-91-2
  • Molecular Weight : 241.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-3436

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-3436
Availability details for
Quantity
  • Availability
  • Add to Cart