COMH4211DF32 Combi-Blocks, Inc.

7-tert-Butyl-4-chloro-5,6,7,8-tetrahydro-benzo[4,5]thieno[2,3-d]pyrimidine

  • MDL number MFCD02994402
  • Cas Number : 381200-95-9
  • Molecular Weight : 280.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-3552

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-3552
Availability details for
Quantity
  • Availability
  • Add to Cart