COMH4211DF35 Combi-Blocks, Inc.

7-(Trifluoromethyl)-1h-indazol-3-amine

  • MDL number MFCD03426698
  • Cas Number : 60330-35-0
  • Molecular Weight : 201.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-3587

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-3587
Availability details for
Quantity
  • Availability
  • Add to Cart