COMH4211DF41 Combi-Blocks, Inc.

5-[2-(Methylthio)pyrimidin-4-yl]thiophene-2-sulfonyl chloride

  • MDL number MFCD00178756
  • Cas Number : 175202-76-3
  • Molecular Weight : 306.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-3655

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-3655
Availability details for
Quantity
  • Availability
  • Add to Cart