COMH4211DF74 Combi-Blocks, Inc.

4-((Methylsulfonyl)methyl)piperidine

  • MDL number MFCD12197684
  • Cas Number : 290328-53-9
  • Molecular Weight : 177.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-3959

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-3959
Availability details for
Quantity
  • Availability
  • Add to Cart