COMH4211DF7E Combi-Blocks, Inc.

2-Isopropyl-6-nitrophenol

  • MDL number MFCD00085099
  • Cas Number : 7545-71-3
  • Molecular Weight : 181.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-4033

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-4033
Availability details for
Quantity
  • Availability
  • Add to Cart