COMH4211DF87 Combi-Blocks, Inc.

N1-[5-(Trifluoromethyl)pyridin-2-yl]ethane-1,2-diamine

  • MDL number MFCD00816592
  • Cas Number : 207557-34-4
  • Molecular Weight : 205.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-4126

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-4126
Availability details for
Quantity
  • Availability
  • Add to Cart