COMH4211DFCD Combi-Blocks, Inc.

[(4-Bromobenzyl)sulfonyl]acetic acid

  • MDL number MFCD09934120
  • Cas Number : 81431-45-0
  • Molecular Weight : 293.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-4731

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-4731
Availability details for
Quantity
  • Availability
  • Add to Cart