COMH4211DFFD Combi-Blocks, Inc.

Bis-(n-boc-aminoxy)-peg3

  • MDL number MFCD28950775
  • Cas Number : 1807518-66-6
  • Molecular Weight : 424.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-5080

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-5080
Availability details for
Quantity
  • Availability
  • Add to Cart