COMH4211E067 Combi-Blocks, Inc.

5-Amino-3-[(4-chlorophenyl)amino]-1h-1,2,4-triazole

  • MDL number MFCD00104001
  • Cas Number : 16650-53-6
  • Molecular Weight : 209.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-5895

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-5895
Availability details for
Quantity
  • Availability
  • Add to Cart