COMH4211E09C Combi-Blocks, Inc.

Boc-3-exo-aminobicyclo[2.2.1]-hept-5-ene-2-exo-carboxylic acid

  • MDL number MFCD02682617
  • Cas Number : 148257-06-1
  • Molecular Weight : 253.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-6303

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-6303
Availability details for
Quantity
  • Availability
  • Add to Cart