COMH4211E0CF Combi-Blocks, Inc.

(S)-5,8A-Dimethyl-3,4,8,8a-tetrahydro-2h,7h-naphthalene-1,6-dione

  • MDL number MFCD08751318
  • Cas Number : 41722-49-0
  • Molecular Weight : 192.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-6669

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-6669
Availability details for
Quantity
  • Availability
  • Add to Cart