COMH4211E0E8 Combi-Blocks, Inc.

6-Chloro-3-indoxyl-alpha-d-glucopyranoside

  • MDL number MFCD04972053
  • Cas Number : 467214-46-6
  • Molecular Weight : 329.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-6843

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-6843
Availability details for
Quantity
  • Availability
  • Add to Cart