COMH4211E0FE Combi-Blocks, Inc.

Caffeic acid

  • MDL number MFCD01325311
  • Cas Number : 331-39-5
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-7043

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-7043
Availability details for
Quantity
  • Availability
  • Add to Cart