COMH4211F0B9 Combi-Blocks, Inc.

Benzyl N-{1-[(4-methoxyphenyl)methyl]-5-oxopyrrolidin-3-yl}carbamate

  • MDL number MFCD32065782
  • Cas Number : 2365419-31-2
  • Molecular Weight : 354.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4883

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4883
Availability details for
Quantity
  • Availability
  • Add to Cart