COMH4211F0BA Combi-Blocks, Inc.

1-(morpholin-4-yl)-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)propan-1-one

  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4893

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4893
Availability details for
Quantity
  • Availability
  • Add to Cart